Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFAT5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | NFAT5 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NFAT5 Polyclonal specifically detects NFAT5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
NFAT5 | |
Polyclonal | |
Rabbit | |
Wnt Signaling Pathway | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
10725 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TNEDMQMQCELFSSPPAVSGNETSTTTTQQVATPGTTMFQTSSSGDGEETGTQAKQIQNSVFQTMVQMQHSGDNQPQVNLFSSTKSMMSVQNSGTQQQGNGLFQQGNEMMSLQSGNFLQQSSHSQAQLFHPQNPIADAQN | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
EC 3.6.1, EC 3.6.3.14, KIAA0827glutamine rich protein H65, NF-AT5NFAT-like protein 1, NFATL1, NFATZ, nuclear factor of activated T-cells 5, tonicity-responsive, OREBP, osmotic response element-binding protein, T-cell transcription factor NFAT5, TonE-binding protein, TonEBP, TONEBPnuclear factor of activated T-cells 5, Tonicity-responsive enhancer-binding protein | |
NFAT5 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title