Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFATC1/NFAT2 Antibody (7A6), Novus Biologicals™
SDP

Catalog No. NB300620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300620 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300620 Supplier Novus Biologicals Supplier No. NB300620
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

NFATC1/NFAT2 Monoclonal specifically detects NFATC1/NFAT2 in Human, Mouse, Rat, Hamster, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Super Shift Assays.

Specifications

Antigen NFATC1/NFAT2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 7A6
Concentration 9.25mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Simple Western, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20 - 1:200, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Gene Accession No. O95644
Gene Alias MGC138448, NFAT transcription complex cytosolic component, NFAT2cytoplasmic 1, NFATC, NF-ATC, NFATc1, NF-ATc1, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1
Gene Symbols NFATC1
Host Species Mouse
Immunogen Bacterially expressed glutathione S-transferase (GST) fusion protein containing NF-ATc1 residues 1-654.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4772
Test Specificity Detects cytosolic nuclear factor of activated T-cells (NFAT 2) from transfected human, mouse, and monkey cells. This does not cross react with NFATp.
Target Species Human, Mouse, Rat, Hamster, Primate, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.