Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFATC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579730
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579730 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579730 Supplier Invitrogen™ Supplier No. PA579730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: mouse thymus tissue, human 22RV1 whole cell. Flow: Hela cell, THP-1 cell, MCF-7 cell, Jurkat cell.

The product of this gene is a component of the nuclear factor of activated T cells DNA-binding transcription complex. This complex consists of at least two components: a preexisting cytosolic component that translocates to the nucleus upon T cell receptor (TCR) stimulation, and an inducible nuclear component. Proteins belonging to this family of transcription factors play a central role in inducible gene transcription during immune response. The product of this gene is an inducible nuclear component. It functions as a major molecular target for the immunosuppressive drugs such as cyclosporin A. Five transcript variants encoding distinct isoforms have been identified for this gene. Different isoforms of this protein may regulate inducible expression of different cytokine genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFATC1
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.9mg NaCl, 0.2 Na2PO4, 4mg trehalose and no preservative
Gene NFATC1
Gene Accession No. O95644
Gene Alias 2210017P03Rik; AI449492; AV076380; MGC138448; NFAT tra; NFAT trans; NFAT transcription complex cytosolic component; NFAT2; NFATc; NF-ATC; NFATC1; NF-ATc1; NF-ATc1.2; Nfatcb; nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 1; nuclear factor of activated T-cells 1; nuclear factor of activated T-cells 'c'; nuclear factor of activated T-cells, cytoplasmic 1; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1; transcription factor NF-ATc
Gene Symbols NFATC1
Host Species Rabbit
Immunogen E. coli-derived human NFAT2 recombinant protein (Position: Q589-K652).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4772
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.