Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFATC1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA123423
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA123423 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA123423 Supplier Thermo Scientific Supplier No. PA123423

Rabbit Polyclonal Antibody

PA1-23423 detects NFATc1 in human samples.

The product of this gene is a component of the nuclear factor of activated T cells DNA-binding transcription complex. This complex consists of at least two components: a preexisting cytosolic component that translocates to the nucleus upon T cell receptor (TCR) stimulation, and an inducible nuclear component. Proteins belonging to this family of transcription factors play a central role in inducible gene transcription during immune response. The product of this gene is an inducible nuclear component. It functions as a major molecular target for the immunosuppressive drugs such as cyclosporin A. Five transcript variants encoding distinct isoforms have been identified for this gene. Different isoforms of this protein may regulate inducible expression of different cytokine genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFATC1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NFATC1
Gene Accession No. O95644
Gene Alias 2210017P03Rik; AI449492; AV076380; MGC138448; NFAT tra; NFAT trans; NFAT transcription complex cytosolic component; NFAT2; NFATc; NF-ATC; NFATC1; NF-ATc1; NF-ATc1.2; Nfatcb; nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 1; nuclear factor of activated T-cells 1; nuclear factor of activated T-cells 'c'; nuclear factor of activated T-cells, cytoplasmic 1; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1; transcription factor NF-ATc
Gene Symbols NFATC1
Host Species Rabbit
Immunogen Synthetic peptide corresponding top residues L(102) S S G H T R P D G A P A(114) of human NFATc1.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4772
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.