Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFATC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14198
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14198 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14198 Supplier Invitrogen™ Supplier No. PA14198
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4198 detects NFAT2 in human samples. NFATc2 is also known as NFAT1 or NFATp. PA1-4198 has been successfully used in Western blot procedures. The PA1-4198 immunogen is a synthetic peptide corresponding to residues A(269) S P Q R S R S P S P Q P(281) of human NFATc2.

The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFATC2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NFATC2
Gene Accession No. Q13469
Gene Alias AI607462; IB-IXL; KIAA0611; NFAT pre-existing s; NFAT pre-existing subunit; NFAT transcription complex, preexisting component; Nfat1; NFAT1-D; Nfatc2; NF-ATc2; Nfatp; NF-ATp; nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 2; nuclear factor of activated T-cells 2; nuclear factor of activated T-cells, cytoplasmic 2; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2; nuclear factor of activated T-cells, preexisting component; OTTHUMP00000031291; preexisting nuclear factor of activated T-cells 2; T cell transcription factor NFAT1; T-cell transcription factor NFAT1
Gene Symbols NFATC2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues A(269) S P Q R S R S P S P Q P(281) of human NFATc2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4773
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.