Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NFATC2IP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565805
Description
Immunogen sequence: SPWKTKLRTKD KEEKKKTEFL DLDNSPLSPP SPRTKSRTHT RALKKLSEVN KRLQDLRSCL SPKPPQGQEQ QGQEDEVVLV E Highest antigen sequence identity to the following orthologs - mouse 67%, rat 69%.
In T-helper 2 (Th2) cells, regulates the magnitude of NFAT-driven transcription of a specific subset of cytokine genes, including IL3, IL4, IL5 and IL13, but not IL2. Recruits PRMT1 to the IL4 promoter; this leads to enhancement of histone H4 'Arg-3'-methylation and facilitates subsequent histone acetylation at the IL4 locus, thus promotes robust cytokine expression. Down-regulates formation of poly-SUMO chains by UBE2I/UBC9.
Specifications
NFATC2IP | |
Polyclonal | |
Unconjugated | |
NFATC2IP | |
45 kDa NF-AT-interacting protein; 45 kDa NFAT-interacting protein; ESC2; NFATC2-interacting protein; NFATC2IP; NIP45; nuclear factor of activated T-cells 2 interacting protein; nuclear factor of activated T-cells, cytoplasmic 2-interacting protein; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2 interacting protein; RAD60 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84901 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NCF5 | |
NFATC2IP | |
Recombinant Human NFATC2IP. Recombinant protein control fragment (Product #RP-106479). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction