Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFATC3/NFAT4 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204026G
Description
NFATC3/NFAT4 Polyclonal antibody specifically detects NFATC3/NFAT4 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
NFATC3/NFAT4 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
NF-AT4, NFAT4nuclear factor of activated T-cells, cytoplasmic 3, NFATc3, NF-ATc3, NFATx, nuclear factor of activated T-cells c3 isoform IE-Xa, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 3, T cell transcription factor NFAT4, T-cell transcription factor NFAT4 | |
The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human Nuclear Factor of Activated T-cells, Cytoplasmic, Calcineurin-dependent 3 using the numbering given in entry NP_775188.1 (GeneID 4775). | |
0.1 mL | |
Adaptive Immunity, Immunology, Wnt Signaling Pathway | |
4775 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction