Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFATC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579734
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579734 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579734 Supplier Invitrogen™ Supplier No. PA579734
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human 22RV1 whole cell. Flow: A431 cell, U20S cell, K562 cell, SiHa cell.

This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein is located only in the cytoplasm. When translation ends, it interacts with the protein that is a functional homolog of yeast Upf2p to trigger mRNA decapping. Use of multiple polyadenylation sites has been noted for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFATC3
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation Antibody with 0.2mg Na2PO4, 0.9mg NaCl, 4mg trehalose and 0.05mg sodium azide
Gene NFATC3
Gene Accession No. P97305, Q12968
Gene Alias C80703; D8Ertd281e; NFAC3; NFAT4; NF-AT4; NFATC3; NF-ATc3; NFATx; NFC3; nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 3; nuclear factor of activated T-cells 3; nuclear factor of activated T-cells c3 isoform IE-Xa; nuclear factor of activated T-cells, cytoplasmic 3; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 3; T cell transcription factor NFAT4; T-cell transcription factor NFAT4
Gene Symbols NFATC3
Host Species Rabbit
Immunogen E. coli-derived human NFAT4 recombinant protein (Position: K630-L712).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18021, 361400, 4775
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.