Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFATC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585559
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585559 Supplier Invitrogen™ Supplier No. PA585559
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (91%), Rat (91%). Positive Control: NFATC4-transfected 293T cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This protein is an adapter or scaffold protein, which participates in the assembly of numerous protein complexes and is involved in several cellular processes such as cell fate determination, cytoskeletal organization, repression of gene transcription, mitosis, cell-cell adhesion, cell differentiation, proliferation and migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFATC4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Nfatc4
Gene Accession No. Q14934
Gene Alias 3110041H08Rik; AW107667; AW546455; Nfat3; NF-AT3; Nfatc4; NF-ATc4; nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 4; nuclear factor of activated T-cells 4; nuclear factor of activated T-cells, cytoplasmic 4; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 4; T-cell transcription factor NFAT3; transcription complex subunit NF-ATc4
Gene Symbols Nfatc4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human NFATC4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4776
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.