Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551659 Supplier Invitrogen™ Supplier No. PA551659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SRNRVIQLST SELGEMELTW QEIMSITELQ GLNAPSEPSF EPQAPAPYLG PPPPTTYCPC SIHPDSGFPL PPPPYELPAS TSHVPDPPYS YGNMAIPVSK PLSLSGLLSE PLQDPLALLD IGLPAGPPKP QEDPES Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%.

Component of the NF-E2 complex essential for regulating erythroid and megakaryocytic maturation and differentiation. Binds to the hypersensitive site 2 (HS2) of the beta-globin control region (LCR). This subunit (NFE2) recognizes the TCAT/C sequence of the AP-1-like core palindrome present in a number of erythroid and megakaryocytic gene promoters. Requires MAFK or other small MAF proteins for binding to the NF-E2 motif. May play a role in all aspects of hemoglobin production from globin and heme synthesis to procurement of iron.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFE2
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NFE2
Gene Accession No. Q16621
Gene Alias leucine zipper protein NF-E2; NFE2; NF-E2; NF-E2/P45; nuclear factor (erythroid-derived 2), 45kD; nuclear factor (erythroid-derived 2), 45kDa; nuclear factor, erythroid 2; nuclear factor, erythroid derived 2; nuclear factor, erythroid-derived 2 45 kDa subunit; p45; p45 NF-E2; p45NFE2; p45nf-e2; Transcription factor NF-E2 45 kDa subunit
Gene Symbols NFE2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NFE2. Recombinant protein control fragment (Product #RP-100710).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4778
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.