Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NFE2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567842
Description
Predicted to react with Mouse and Rat samples.
Component of the NF-E2 complex essential for regulating erythroid and megakaryocytic maturation and differentiation. Binds to the hypersensitive site 2 (HS2) of the beta-globin control region (LCR). This subunit (NFE2) recognizes the TCAT/C sequence of the AP-1-like core palindrome present in a number of erythroid and megakaryocytic gene promoters. Requires MAFK or other small MAF proteins for binding to the NF-E2 motif. May play a role in all aspects of hemoglobin production from globin and heme synthesis to procurement of iron.
Specifications
NFE2 | |
Polyclonal | |
Unconjugated | |
NFE2 | |
leucine zipper protein NF-E2; NFE2; NF-E2; NF-E2/P45; nuclear factor (erythroid-derived 2), 45kD; nuclear factor (erythroid-derived 2), 45kDa; nuclear factor, erythroid 2; nuclear factor, erythroid derived 2; nuclear factor, erythroid-derived 2 45 kDa subunit; p45; p45 NF-E2; p45NFE2; p45nf-e2; Transcription factor NF-E2 45 kDa subunit | |
Rabbit | |
affinity chromatography | |
RUO | |
18022, 366998, 4778 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q07279, Q16621, Q6AYT2 | |
NFE2 | |
Synthesized peptide derived from human NFE2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction