Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFE2L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568511
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568511 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568511 Supplier Invitrogen™ Supplier No. PA568511
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%.

This gene encodes a protein that is involved in globin gene expression in erythrocytes. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene, NFE2L1, and for 'nuclear respiratory factor 1' which has an official symbol of NRF1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFE2L1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NFE2L1
Gene Accession No. Q61985
Gene Alias AA408798; AW212678; bZIP protein; endoplasmic reticulum membrane sensor NFE2L1; HBZ17; Lcrf1; LCR-F1; locus control region-factor 1; Nfe2l1; NF-E2-related factor 1; NFE2-related factor 1; Nrf1; nuclear factor (erythroid-derived 2)-like 1; nuclear factor erythroid 2-like 1; nuclear factor erythroid 2-related factor 1; nuclear factor, erythroid 2 like 1; nuclear factor, erythroid 2-like 1; nuclear factor, erythroid derived 2, like 1; nuclear factor, erythroid derived 2,-like 1; Protein NRF1, p110 form; Protein NRF1, p120 form; TCF11; TCF-11; transc; Transcription factor 11; transcription factor 11 (basic leucine zipper type); transcription factor HBZ17; transcription factor LCR-F1; Transcription factor NRF1
Gene Symbols NFE2L1
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse NFE2L1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18023
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.