Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFE2L3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102015
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102015 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102015 Supplier Invitrogen™ Supplier No. PA5102015
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NFE2L3.

NFE2L3 activates erythroid-specific, globin gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFE2L3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NFE2L3
Gene Accession No. Q9Y4A8
Gene Alias LOW QUALITY PROTEIN: nuclear factor erythroid 2-related factor 3; Nfe2l3; NF-E2-related factor 3; NFE2-related factor 3; Nrf3; nuclear factor (erythroid-derived 2)-like 3; nuclear factor erythroid 2-like 3; nuclear factor erythroid 2-related factor 3; nuclear factor, erythroid 2 like 3; nuclear factor, erythroid 2-like 3; nuclear factor, erythroid derived 2, like 3; nuclear factor-erythroid 2 p45-related factor 3; nuclear factor-erythroid 2-related factor 3
Gene Symbols NFE2L3
Host Species Rabbit
Immunogen A synthesized peptide derived from human NFE2L3(Accession Q9Y4A8), corresponding to amino acid residues A630-V680.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9603
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.