Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFE2L3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599083
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599083 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599083 Supplier Invitrogen™ Supplier No. PA599083
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NFE2L3 Polyclonal Antibody for ELISA, IHC (P), Western Blot

NFE2L3 activates erythroid-specific, globin gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFE2L3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NFE2L3
Gene Accession No. Q9Y4A8
Gene Alias LOW QUALITY PROTEIN: nuclear factor erythroid 2-related factor 3; Nfe2l3; NF-E2-related factor 3; NFE2-related factor 3; Nrf3; nuclear factor (erythroid-derived 2)-like 3; nuclear factor erythroid 2-like 3; nuclear factor erythroid 2-related factor 3; nuclear factor, erythroid 2 like 3; nuclear factor, erythroid 2-like 3; nuclear factor, erythroid derived 2, like 3; nuclear factor-erythroid 2 p45-related factor 3; nuclear factor-erythroid 2-related factor 3
Gene Symbols NFE2L3
Host Species Rabbit
Immunogen Recombinant Human Nuclear factor erythroid 2-related factor 3 protein (132-260AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9603
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.