Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFIA Antibody (PCRP-NFIA-2C6), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314016MFV610
Description
NFIA Monoclonal antibody specifically detects NFIA in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
NFIA | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
PCRP-NFIA-2C6 | |
Flow Cytometry | |
CCAAT-box-binding transcription factor, CTF, DKFZp686J23256, FLJ39164, KIAA1439DKFZp434L0422, NF1-A, NF-I/A, NFI-A, NFI-L, nuclear factor 1 A-type, Nuclear factor 1/A, nuclear factor I/ATGGCA-binding protein | |
Recombinant fragment of human NFIA protein corresponding to protein domain (Uniprot: Q15784) | |
0.1 mL | |
Cell Cycle and Replication | |
4774 | |
Store at 4C in the dark. | |
IgG2c κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction