Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFIA Monoclonal Antibody (16H11)
GREENER_CHOICE

Catalog No. PIMA549256 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549256 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549256 Supplier Invitrogen™ Supplier No. MA549256
  • $481.00
    $425.65 / Each of 1

    Save $55.35

    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Immunogen sequence is different from the related mouse sequence by one amino acid, and identical to the related rat sequence. Positive Control - WB: human Hela whole cell, human HEK293 whole cell. IHC: human intestinal cancer tissue, human intestinal cancer tissue, human tonsil tissue. ICC/IF: A431 cell. Flow: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

NF-1, also designated CTF, consists of a family of CCAAT box binding proteins that stimulate DNA replication and activate transcription. Analysis of human NF-1 messenger RNA has revealed two forms of the NF-1 protein arising from an alternate splicing of a single NF-1 gene. NF-1 binds its consensus DNA element as a homodimer via an amino-terminal DNA binding domain, and activates transcription through a putatively novel, proline-rich, carboxy terminal transactivation domain. The NF-1 protein has been shown to recognize and bind the adenovirus type 2 promoter and activate transcription of herpes simplex virus thymidine kinase genes. The NF-1 consensus element has been found in the upstream promoter region of myriad eukaryotic genes, including that of Ha-Ras, alpha-globin, HSP 70, GRP 78, Histone H1, myelin basic protein and in the Xenopus laevis vitellogenin gene promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFIA
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 16H11
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene NFIA
Gene Accession No. Q12857
Gene Alias 1110047K16Rik; 9430022M17Rik; CCAAT-box-binding transcription factor; CTF; KIAA1439; NF1A; NF1-A; Nf1l21; NF-I/A; Nfia; NFI-A; NFI-L; Nuclear factor 1 A-type; nuclear factor 1/A; nuclear factor I A; nuclear factor I/A; TGGCA-binding protein; Unknown (protein for MGC:128467)
Gene Symbols NFIA
Host Species Mouse
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human NFIA (180-224aa AYFVHAADSSQSESPSQPSDADIKDQPENGHLGFQDSFVTSGVFS).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4774
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.