Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFIA Monoclonal Antibody (OTI4G7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA811198

Description
NF-1, also designated CTF, consists of a family of CCAAT box binding proteins that stimulate DNA replication and activate transcription. Analysis of human NF-1 messenger RNA has revealed two forms of the NF-1 protein arising from an alternate splicing of a single NF-1 gene. NF-1 binds its consensus DNA element as a homodimer via an amino-terminal DNA binding domain, and activates transcription through a putatively novel, proline-rich, carboxy terminal transactivation domain. The NF-1 protein has been shown to recognize and bind the adenovirus type 2 promoter and activate transcription of herpes simplex virus thymidine kinase genes. The NF-1 consensus element has been found in the upstream promoter region of myriad eukaryotic genes, including that of Ha-Ras, alpha-globin, HSP 70, GRP 78, Histone H1, myelin basic protein and in the Xenopus laevis vitellogenin gene promoter.Specifications
NFIA | |
Monoclonal | |
Unconjugated | |
NFIA | |
BRMUTD, CTF, NF-I/A, NF1-A, NFI-A, NFI-L | |
Mouse | |
Affinity Chromatography | |
RUO | |
4774 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI4G7 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q12857 | |
NFIA | |
Human recombit protein fragment corresponding to amino acids 200-509 of human NFIA produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction