Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFIB Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204121AF532
Description
NFIB Polyclonal antibody specifically detects NFIB in Human samples. It is validated for Western BlotSpecifications
| NFIB | |
| Polyclonal | |
| Western Blot | |
| CCAAT-box-binding transcription factor, CTF, HMGIC/NFIB, NF-I/B, NFI-B, NFIB2, NFIB3, NFI-RED, nuclear factor 1 B-type, Nuclear factor 1/B, nuclear factor I/BNF1-B, TGGCA-binding protein | |
| The immunogen recognized by this antibody maps to a region between residue 225 and 275 of human Nuclear Factor I/B using the numbering given in entry NP_005587.2 (GeneID 4781). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 4781 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction