Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFIC Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171879JF646
Description
NFIC Polyclonal antibody specifically detects NFIC in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
NFIC | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
CCAAT-binding transcription factor, CTF5, CTFCCAAT-box-binding transcription factor, MGC20153, NF1-C, NFI, NF-I, NF-I/C, NFI-C, nuclear factor 1 C-type, Nuclear factor 1/C, Nuclear factor I/C, nuclear factor I/C (CCAAT-binding transcription factor), TGGCA-binding protein | |
The immunogen for this product maps to a region between residue 250 and 300 of human Nuclear Factor I/C using the numbering given in entry EAW69324.1 (GeneID 4782). | |
0.1 mL | |
Cell Cycle and Replication | |
4782 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction