Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFIC Monoclonal Antibody (1D6)
GREENER_CHOICE

Catalog No. PIMA519452
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA519452 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA519452 Supplier Invitrogen™ Supplier No. MA519452
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: TEDMEGGISS PVKKTEMDKS PFNSPSPQDS PRLSSFTQHH RPVIAVHSGI ARSPHPSSAL HFPTTSILPQ TASTYFPHTA IRYPPHLNPQ DPLKDLVSLA CDPASQQPGP.

Recognizes and binds the palindromic sequence 5'-TTGGCNNNNNGCCAA-3' present in viral and cellular promoters and in the origin of replication of adenovirus type 2. These proteins are individually capable of activating transcription and replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFIC
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1D6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene NFIC
Gene Accession No. P08651
Gene Alias 1110019L22Rik; 1500041O16Rik; AA589446; AI746521; CCAAT-box-binding transcription factor; CTF; CTF5; KIAA1439; NF1-A; NF1-B; NF1-C; NF1-X; NFI; NF-I; NF-I/A; NF-I/B; NF-I/C; NF-I/X; NFIA; NFI-A; NFIB; NFI-B; Nfic; NFI-C; NFIX; NFI-X; Nuclear factor 1 C-type; Nuclear factor 1/A; Nuclear factor 1/B; nuclear factor 1/C; Nuclear factor 1/X; nuclear factor 1-C2; nuclear factor I C; Nuclear factor I/A; Nuclear factor I/B; nuclear factor I/C; nuclear factor I/C (CCAAT-binding transcription factor); Nuclear factor I/X; olfactory epithelium nuclear factor I-C; TGGCA-binding protein
Gene Symbols NFIC
Host Species Mouse
Immunogen NFIC (NP_005588, 314 a.a. ~ 423 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4782
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.