Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFIC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590204
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590204 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590204 Supplier Invitrogen™ Supplier No. PA590204
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MYSSPLCLTQ DEFHPFIEAL LPHVRAFAYT WFNLQARKRK YFKKHEKRMS KDEERAVKDE LLGEKPEVKQ KWASRLLAKL RKDIRPECRE DFVLSITGKK APGCVLSNPD QKGKMRRIDC LRQADKVWRL DLVMVILFKG IPLESTDGER LVKAAQCGHP VLCVQPHHIG VAVKELDLYL AYFVRERDAE QSGSPRTGMG SDQEDSKPIT LDTTDFQESF; Positive Samples: HeLa, LO2, MCF-7, DU145, NIH/3T3, mouse liver, mouse skeletal muscle, rat liver; Cellular Location: Nucleus.

Recognizes and binds the palindromic sequence 5'-TTGGCNNNNNGCCAA-3' present in viral and cellular promoters and in the origin of replication of adenovirus type 2. These proteins are individually capable of activating transcription and replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFIC
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NFIC
Gene Accession No. P08651, P70255
Gene Alias 1110019L22Rik; 1500041O16Rik; AA589446; AI746521; CCAAT-box-binding transcription factor; CTF; CTF5; KIAA1439; NF1-A; NF1-B; NF1-C; NF1-X; NFI; NF-I; NF-I/A; NF-I/B; NF-I/C; NF-I/X; NFIA; NFI-A; NFIB; NFI-B; Nfic; NFI-C; NFIX; NFI-X; Nuclear factor 1 C-type; Nuclear factor 1/A; Nuclear factor 1/B; nuclear factor 1/C; Nuclear factor 1/X; nuclear factor 1-C2; nuclear factor I C; Nuclear factor I/A; Nuclear factor I/B; nuclear factor I/C; nuclear factor I/C (CCAAT-binding transcription factor); Nuclear factor I/X; olfactory epithelium nuclear factor I-C; TGGCA-binding protein
Gene Symbols NFIC
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human NFIC (NP_0055882).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18029, 29228, 4782
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.