Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFIC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530101 Supplier Invitrogen™ Supplier No. PA530101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549. Predicted reactivity: Mouse (96%), Rat (96%), Xenopus laevis (83%), Chicken (89%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Recognizes and binds the palindromic sequence 5'-TTGGCNNNNNGCCAA-3' present in viral and cellular promoters and in the origin of replication of adenovirus type 2. These proteins are individually capable of activating transcription and replication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFIC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene NFIC
Gene Accession No. P08651
Gene Alias 1110019L22Rik; 1500041O16Rik; AA589446; AI746521; CCAAT-box-binding transcription factor; CTF; CTF5; KIAA1439; NF1-A; NF1-B; NF1-C; NF1-X; NFI; NF-I; NF-I/A; NF-I/B; NF-I/C; NF-I/X; NFIA; NFI-A; NFIB; NFI-B; Nfic; NFI-C; NFIX; NFI-X; Nuclear factor 1 C-type; Nuclear factor 1/A; Nuclear factor 1/B; nuclear factor 1/C; Nuclear factor 1/X; nuclear factor 1-C2; nuclear factor I C; Nuclear factor I/A; Nuclear factor I/B; nuclear factor I/C; nuclear factor I/C (CCAAT-binding transcription factor); Nuclear factor I/X; olfactory epithelium nuclear factor I-C; TGGCA-binding protein
Gene Symbols NFIC
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NFIC. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4782
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.