Learn More
NFIC Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303123AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
CCAAT-binding transcription factor,nuclear factor I/CSpecifications
NFIC | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P08651, P70255 | |
Nfic | |
Between 200 and 250 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
NFIC | |
CTF, CTF5, NF-I, NFI, CCAAT-box-binding transcription factor, NF-I/C, NF1-C, TGGCA-binding protein, nuclear factor 1 C-type | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18029, 4782 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.