Learn More
NFIC Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303124A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
CCAAT-binding transcription factor,nuclear factor I/CSpecifications
NFIC | |
Polyclonal | |
Unconjugated | |
NFIC | |
CTF, CTF5, NF-I, NFI, CCAAT-box-binding transcription factor, NF-I/C, NF1-C, TGGCA-binding protein, nuclear factor 1 C-type | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4782 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P08651 | |
NFIC | |
Between 250 and 300 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.