Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFIL3 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP141077AF405
Description
NFIL3 Polyclonal antibody specifically detects NFIL3 in Human samples. It is validated for Western BlotSpecifications
NFIL3 | |
Polyclonal | |
Western Blot | |
E4BP4IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A | |
The immunogen for this product maps to a region between residue 412 and 462 of human E4 promoter binding-protein 4 using the numbering given in entry NP_005375.2 (GeneID 4783). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
4783 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction