Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFIL3 Mouse anti-Human, Porcine, Clone: 2F4B4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | NFIL3 |
---|---|
Clone | 2F4B4 |
Concentration | 0.93 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6798
![]() ![]() |
Proteintech Group Inc
663681IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
MCM2 (Mini chromosome maintece protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. Mincheva et al. (1994) mapped the gene to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).Specifications
NFIL3 | |
0.93 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
E4 promoter binding protein 4, E4BP4, IL3BP1, NF IL3A, NFIL3, NFIL3A | |
NFIL3 | |
IgG1 | |
Protein G | |
Antibody |
2F4B4 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig | |
Q16649 | |
100153822, 4783 | |
NFIL3 Fusion Protein Ag2380 | |
Primary | |
-20°C | |
NFIL3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title