Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFIL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517978
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517978 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517978 Supplier Invitrogen™ Supplier No. PA517978
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.

Expression of interleukin-3 (IL3; MIM 147740) is restricted to activated T cells, natural killer (NK) cells, and mast cell lines. Transcription initiation depends on the activating capacity of specific protein factors, such as NFIL3, that bind to regulatory regions of the gene, usually upstream of the transcription start site (Zhang et al., 1995 [PubMed 7565758]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFIL3
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene NFIL3
Gene Accession No. Q16649
Gene Alias AV225605; E4 promoter binding-protein 4; E4 promoter-binding protein 4; E4BP4; Embryo implantation-related NFIL3/E4BP4-like transcription factor; IL3BP1; Interleukin-3 promoter transcriptional activator; interleukin-3-binding protein 1; NF IL3A; NFIL3; NFIL3A; NF-IL3A; nuclear factor interleukin 3 regulated protein; nuclear factor interleukin-3-regulated protein; nuclear factor, interleukin 3 regulated; nuclear factor, interleukin 3, regulated; OTTHUMP00000021633; transcriptional activator NF-IL3A
Gene Symbols NFIL3
Host Species Goat
Immunogen Synthetic peptide sequence (RLIATQPISASDSG) corresponding to the C-terminus amino acids of NFIL3 (aa 449-462).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4783
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.