Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFIL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578077
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578077 Supplier Invitrogen™ Supplier No. PA578077
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: HepG2, HepG2 nuclear extract Predicted Reactivity: Rat (80%), Chicken (89%), Sheep (87%), Rhesus Monkey (98%), Bovine (87%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Expression of interleukin-3 (IL3; MIM 147740) is restricted to activated T cells, natural killer (NK) cells, and mast cell lines. Transcription initiation depends on the activating capacity of specific protein factors, such as NFIL3, that bind to regulatory regions of the gene, usually upstream of the transcription start site (Zhang et al., 1995 [PubMed 7565758]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFIL3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene NFIL3
Gene Accession No. Q16649
Gene Alias AV225605; E4 promoter binding-protein 4; E4 promoter-binding protein 4; E4BP4; Embryo implantation-related NFIL3/E4BP4-like transcription factor; IL3BP1; Interleukin-3 promoter transcriptional activator; interleukin-3-binding protein 1; NF IL3A; NFIL3; NFIL3A; NF-IL3A; nuclear factor interleukin 3 regulated protein; nuclear factor interleukin-3-regulated protein; nuclear factor, interleukin 3 regulated; nuclear factor, interleukin 3, regulated; OTTHUMP00000021633; transcriptional activator NF-IL3A
Gene Symbols NFIL3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NFIL3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4783
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.