Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFIL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110026
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110026 Supplier Invitrogen™ Supplier No. PA5110026
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AYAQEIQKLS NSTAVYFQDY QTSKSNVSSF VDEHEPSMVS SSCISVIKHS PQSSLSDVSE VSSVEHTQES SVQGSCRSPE NKFQIIKQEP MELESYTREP RDDRGSYTAS IYQNYMGNSF SGYSHSPPLL QVNRSSSNSP RTSETDDGVV GKSSDGEDEQ QVPKGPIHSP VELKHVHATV VKVPEVNSSA LPHKLRIKAK AMQIKVEAFD NEFEATQKLS SPIDMTSKRH FELEKHSAPS MVHSSLTPFS VQVTNIQDWS LKSEHWHQKE LSGKTQNSFK TGVVEMKDSG YKVSDPENLY LKQGIANLSA EVVSLKRLIA TQPISASDSG.

Expression of interleukin-3 (IL3; MIM 147740) is restricted to activated T cells, natural killer (NK) cells, and mast cell lines. Transcription initiation depends on the activating capacity of specific protein factors, such as NFIL3, that bind to regulatory regions of the gene, usually upstream of the transcription start site (Zhang et al., 1995 [PubMed 7565758]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFIL3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NFIL3
Gene Accession No. Q16649
Gene Alias AV225605; E4 promoter binding-protein 4; E4 promoter-binding protein 4; E4BP4; Embryo implantation-related NFIL3/E4BP4-like transcription factor; IL3BP1; Interleukin-3 promoter transcriptional activator; interleukin-3-binding protein 1; NF IL3A; NFIL3; NFIL3A; NF-IL3A; nuclear factor interleukin 3 regulated protein; nuclear factor interleukin-3-regulated protein; nuclear factor, interleukin 3 regulated; nuclear factor, interleukin 3, regulated; OTTHUMP00000021633; transcriptional activator NF-IL3A
Gene Symbols NFIL3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 133-462 of human NFIL3 (NP_005375.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4783
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.