Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFIX Antibody (HL1269) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325576
Description
NFIX Monoclonal antibody specifically detects NFIX in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
NFIX | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
HL1269 | |
Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 1:100-1:1000 | |
CTF, NF1ACCAAT-box-binding transcription factor, NF1-X, NF-I/X, NFI-X, nuclear factor 1 X-type, Nuclear factor 1/X, Nuclear factor I/X, nuclear factor I/X (CCAAT-binding transcription factor), TGGCA-binding protein | |
Recombinant protein encompassing a sequence within the center region of human NFIX. The exact sequence is proprietary. | |
100 μL | |
Cell Cycle and Replication | |
4784 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction