Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFIX Recombinant Rabbit Monoclonal Antibody (HL1269)
GREENER_CHOICE

Catalog No. PIMA546983
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546983 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546983 Supplier Invitrogen™ Supplier No. MA546983
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

NF-1, also designated CTF, consists of a family of CCAAT box binding proteins that stimulate DNA replication and activate transcription. Analysis of human NF-1 messenger RNA has revealed two forms of the NF-1 protein arising from an alternate splicing of a single NF-1 gene. NF-1 binds its consensus DNA element as a homodimer via an amino-terminal DNA binding domain, and activates transcription through a putatively novel, proline-rich, carboxy terminal transactivation domain. The NF-1 protein has been shown to recognize and bind the adenovirus type 2 promoter and activate transcription of herpes simplex virus thymidine kinase genes. The NF-1 consensus element has been found in the upstream promoter region of myriad eukaryotic genes, including that of Ha-Ras, alpha-globin, HSP 70, GRP 78, Histone H1, myelin basic protein and in the Xenopus laevis vitellogenin gene promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFIX
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1269
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene NFIX
Gene Accession No. P70257, Q14938
Gene Alias CCAAT-binding transcription factor; CCAAT-box-binding transcription factor; CTF; KIAA1439; MRSHSS; NF1A; NF1-A; NF1-B; NF1-C; NF1-X; NF1-X1 protein; NFI; NF-I/A; NF-I/B; NF-I/C; NF-I/X; NFIA; NFI-A; NFIB; NFI-B; NFIC; NFI-C; NFIX; NFI-X; Nuclear factor 1 X-type; Nuclear factor 1/A; Nuclear factor 1/B; Nuclear factor 1/C; nuclear factor 1/X; nuclear factor I X; Nuclear factor I/A; Nuclear factor I/B; Nuclear factor I/C; nuclear factor I/X; nuclear factor I/X (CCAAT-binding transcription factor); SOTOS2; TGGCA-binding protein
Gene Symbols NFIX
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NFIX. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18032, 4784, 81524
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.