Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFkB p105/p50 Antibody (2J10D7), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056583AF647
Description
NFkB p105/p50 Monoclonal specifically detects NFkB p105/p50 in Human, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
NFkB p105/p50 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Predicted to react with Mouse and Rat which are 86% homologous with the immunogen. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay | |
2J10D7 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated | |
P25942 | |
NFKB1 | |
A portion of amino acids 150-200 of human NF-kB (p50) was used as the immunogen. | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Diabetes Research, DNA Repair, Neurodegeneration, Neuroscience, Nucleotide Excision Repair, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
4790 | |
Human, Rat, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction