Learn More
Description
Specifications
Specifications
| Antigen | NFkB p105/p50 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoassay |
| Classification | Monoclonal |
| Clone | 2J10D7 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2-5 ug/ml, Flow Cytometry, Immunohistochemistry-Paraffin 5 ug/ml, Knockout Validated |
| Gene Accession No. | P25942 |
| Gene Alias | DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50 |
| Gene Symbols | NFKB1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
