Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFkB p105/p50 Antibody (2J10D7), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056583V
Description
NFkB p105/p50 Monoclonal specifically detects NFkB p105/p50 in Human, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
NFkB p105/p50 | |
Monoclonal | |
DyLight 405 | |
P25942 | |
NFKB1 | |
A portion of amino acids 150-200 of human NF-kB (p50) was used as the immunogen. | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Diabetes Research, DNA Repair, Neurodegeneration, Neuroscience, Nucleotide Excision Repair, Signal Transduction, Transcription Factors and Regulators | |
4790 | |
Human, Rat, Rabbit | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay | |
2J10D7 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated | |
DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Cross reacts with Human. Predicted to react with Mouse and Rat which are 86% homologous with the immunogen. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction