Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ NFkB p50 (NLS) Inhibitor Peptide Set
SDP

Catalog No. NBP229323 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229323 2 mg
NBP2293235 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229323 Supplier Novus Biologicals™ Supplier No. NBP229323
Add to Cart
Add to Cart

Applications: Functional (Inhibition)

Specifications

Host Species Human, Mouse, Rat, Bovine, Canine, Chicken, Xenopus
Components NF-kB p50 (NLS) Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKVQRKRQKLM (p50 NLS sequence: VQRKRQKLM). Molecular weight: 3529. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361
For Use With (Application) Functional (Inhibition)
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Quantity 2 mg
Product Type NFkB p105/p50 Inhibitor Peptide Set
Molecular Weight (g/mol) 3529
Inhibitors NFkB p105/p50
Form Lyophilized

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.