Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFkB p50/p105 Polyclonal Antibody, eBioscience™
GREENER_CHOICE

Catalog No. 5013527
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-135-27 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-135-27 Supplier Invitrogen™ Supplier No. 14673281
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Description: The polyclonal antibody reacts with mouse, rat, and human NFkB p50; the antibody was raised against a peptide mapping to the carboxy terminus of human NFkB p50. Members of the rel/NFkB family of transcription factors are involved in the regulation of cellular responses, such as growth, development, and the inflammatory response. They share a structural motif known as the rel homology region (RHR), the C-terminal one third of which mediates protein dimerization (2, 6, 8). Complexes of p50 (NF-kB1) or p52 (NF-kB2) are generated through the processing of p105 and p100 precursors, respectively. These are usually associated with members of the Rel family (p65, c-Rel, Rel B). The homo- and heterodimer formed through combinations of NFkB/Rel proteins bind distinct kB sites to regulate the transcription of different genes (7, 9). In resting cells, NFkB is retained in the cytoplasm bound to inhibitory proteins of the IkB family. Degradation of IkB proteins occurs with cell activation, via of variety of signals, including inflammatory cytokines and bacterial lipopolysaccharides (LPS) as well as oxidative and fluid mechanical stress. This results in nuclear translocation of NFkB and the transcriptional gene activation of proinflammatory genes (1, 9). It has been suggested that NFkB plays a role in the development of numerous pathological states. Activation of NFkB induces gene programs leading to transcription of factors that promote inflammation, such as leukocyte adhes...

NF-kB is a nuclear transcription factor activated by various extra and intracellular stimuli such as cytokines, UV radiation, stress, in injury, and by bacterial and viral products. It is involved in regulation of various cellular events including cell growth, differentiation, proliferation, apoptosis and inflammation. NFKB1 (p50), a 50KDa functional sub-unit of NF-kB, is a member of Rel protein family. It is synthesized as a p105 precursor protein and consists of an N-terminal conserved RHD-region containing nuclear localization signal, DNA-binding and dimerization domains. NFKB1 (p50) forms homodimers or heterodimerizes with p65, forming the functional NF-kB factor. NFKB1 (p50) directs the nuclear translocation of NF-kB and is instrumental in its DNA-bidning. Pathological role of NF-kB has been suggested in AIDS, hematogenic cancer cell metastasis and rheumatoid arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFkB p50/p105
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with BSA and 0.1% sodium azide
Gene NFKB1
Gene Accession No. P19838, P25799, Q63369
Gene Alias CVID12; DKFZp686C01211; DNA-binding factor KBF1; EBP-1; KBF1; MGC54151; NF kappaB1; NF KB; NFkappaB; NF-kappaB; NF-kappa-B; NF-kappaB p50; NF-kappaB1; NF-kappa-B1 p84/NF-kappa-B1 p98; NF-kappabeta; NF-kB; NF-kB p50/p105; NFKB1; NF-KB1; NFKB-p105; NFKB-p50; nuclear factor; nuclear factor kappa b; nuclear factor kappa B subunit 1; nuclear factor kappa-B DNA binding subunit; nuclear factor kappaB p50; nuclear factor NF-kappa-B p105 subunit; Nuclear factor NF-kappa-B p50 subunit; nuclear factor of kappa light chain gene enhancer in B-cells 1, p105; nuclear factor of kappa light polypeptide gene enhancer in B cells 1, p105; nuclear factor of kappa light polypeptide gene enhancer in B-cells 1; nuclear factor of kappa light polypeptide gene enhancer in B-cells 1, p105; nuclear factor-kappaB p50; p105; p50; p50 subunit of NF kappaB; p50 subunit of NF-kappaB; p50/p105
Gene Symbols NFKB1
Host Species Rabbit
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18033, 4790, 81736
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.