Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NFkB p52/p100 Monoclonal Antibody (E.212.1)
Rabbit Monoclonal Antibody
Supplier: Invitrogen™ MA515110
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other family members.
The NFkB transcription factor was originally identified as a protein complex consisting of a DNA binding subunit and an associated protein. The subunit is functionally related to c-Rel p75 and Rel B p68. The p50 subunit was initially believed to be a functionally unique protein derived from the amino-terminus of a precursor designated p105. A cDNA has been isolated that encodes an alternative DNA binding subunit of NFkB. It is synthesized as a protein that is expressed in a variety of cell types and, like p105, undergoes cleavage to generate its NFkB subunit, in this case a protein designated p52 (previously referred to as p49). In contrast to p50 derived from p105, p52 acts in synergy with p65 to stimulate the HIV enhancer in transiently transfected Jurkat cells.
Specifications
NFkB p52/p100 | |
Monoclonal | |
52 μg/mL | |
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
Q00653 | |
Nfkb2 | |
Synthetic peptide corresponding to residues at the amino-terminus of human NF-kappaB2 p100/p52. | |
100 μL | |
Primary | |
Human, Monkey | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
E.212.1 | |
Unconjugated | |
Nfkb2 | |
CVID10; DNA-binding factor KBF2; H2TF1; Lymphocyte translocation chromosome 10 protein; lyt; LYT10; LYT-10; NF kappaB2; NF KB; NF-kappaB2; nfkb p100; NF-kB p52/p100; NFKB, p52/p100 subunit; NFKB2; NF-kB2; nuclear factor; nuclear factor kappa b; nuclear factor kappa B subunit 2; nuclear factor Kappa-B, subunit 2; nuclear factor NF-kappa-B p100 subunit; Nuclear factor NF-kappa-B p52 subunit; nuclear factor of Kappa light chain gene enhancer in B cells 2; nuclear factor of kappa light polypeptide gene enhancer in B cells 2, p49/p100; Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2; nuclear factor of kappa light polypeptide gene enhancer in B-cells 2 (p49/p100); nuclear factor of kappa light polypeptide gene enhancer in B-cells 2, p49/p100; oncogene Lyt-10; p100; p49; p49/p100; p50B; p52; RGD1307189; transcription factor NFKB2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
4791 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction