Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFKBID Antibody, Novus Biologicals™
SDP

Catalog No. NBP17985620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17985620 20 μL
NBP179856 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17985620 Supplier Novus Biologicals Supplier No. NBP17985620UL

Rabbit Polyclonal Antibody

NFKBID Polyclonal specifically detects NFKBID in Human samples. It is validated for Western Blot.

Specifications

Antigen NFKBID
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_640332
Gene Alias delta, IkappaBdelta, I-kappa-B-delta, IkappaBNSikappaBdelta, ikB-delta, IKBNS, MGC11314, MGC149503, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, TA-NFKBHNF-kappa-B inhibitor delta, T-cell activation NFKB-like protein
Gene Symbols NFKBID
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal of human NFKBIDThe immunogen for this antibody is NFKBID. Peptide sequence QARDRLDCVHMLLQMGANHTSQEIKSNKTVLHLAVQAANPTLVQLLLELP.
Molecular Weight of Antigen 33 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84807
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.