Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFKBID Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598714
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598714 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598714 Supplier Invitrogen™ Supplier No. PA598714
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NFKBID Polyclonal Antibody for Western Blot, ELISA

May regulate the expression of IL-2, IL-6, and other cytokines through regulation on NF-kappa-B activity. Functions in the regulation of inflammatory responses. May also regulate TCR-induced negative selection of thymocytes.I-kappa-BNS, the nuclear I-kappa-B-like protein encoded by Nfkbid, is required for the development of marginal zone and peritoneal B-1 B cells and additionally required for extrafollicular antibody responses to T-independent and -dependent immunogens.I-kappa-BNS has been identified as a member of the I-kappa-B family of NF-kappa-B inhibitors, which undergoes induction upon TCR signaling. I-kappa-BNS mediates both positive and negative gene regulation, depending on individual cell type and/or cytokine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFKBID
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NFKBID
Gene Accession No. Q8NI38
Gene Alias ikappaBdelta; I-kappa-B-delta; IkappaBNS; IkB-delta; IKBNS; NF-kappa B inhibitor; NF-kappa-B inhibitor delta; NFKB inhibitor delta; Nfkbid; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, delta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, delta; TA-NFKBH; T-cell activation NFKB-like protein
Gene Symbols NFKBID
Host Species Rabbit
Immunogen Recombinant Human NF-kappa-B inhibitor delta protein (1-150AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84807
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.