Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFKBIL1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP170344SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP170344SS 0.025 mg
NBP170344 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP170344SS Supplier Novus Biologicals Supplier No. NBP170344SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NFKBIL1 Polyclonal specifically detects NFKBIL1 in Human, Amphibian, Bovine, Canine, Equine, Primate, Chimpanzee samples. It is validated for Western Blot.

Specifications

Antigen NFKBIL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 ug/ml
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. NP_004998.3
Gene Alias IkappaBL, I-kappa-B-like protein, IKBLikappaBL, Inhibitor of kappa B-like protein, LST1, NF-kappa-B inhibitor-like protein 1, nuclear factor of kappa light polypeptide gene enhancer in B-cellsinhibitor-like 1NFKBIL
Gene Symbols NFKBIL1
Host Species Rabbit
Immunogen Amino acids RRYLRVQQVRWHPDRFLQR (331-349) of human NFKBIL1 were used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4795
Test Specificity There are multiple isoforms of this protoein. The amino acid sequenced used as immunogen is provided to help researchers determine suitability of this antibody for their testing.
Target Species Human, Amphibian, Bovine, Canine, Equine, Primate, Chimpanzee
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.