Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFKBIL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568974
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568974 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568974 Supplier Thermo Scientific Supplier No. PA568974
Add to Cart
Add to Cart

NFKBIL1 Polyclonal Antibody

NFKBIL1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen NFKBIL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NFKBIL1
Gene Accession No. O88995
Gene Alias DADB-70P7.2; Def-7; ikappaBL; I-kappa-B-like protein; Ikbl; Inhibitor of kappa B-like protein; LST1; NF-kappa-B inhibitor-like protein 1; NFKB inhibitor like 1; NFKBIL; NFKBIL1; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor like 1; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 1
Gene Symbols NFKBIL1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human NFKBIL1 (aa 320-369).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18038
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.