Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFKBIL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587914 Supplier Invitrogen™ Supplier No. PA587914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSNPSPQVPE EEASTSVCRP KSSMASTSRR QRRERRFRRY LSAGRLVRAQ ALLQRHPGLD VDAGQPPPLH RACARHDAPA LCLLLRLGAD PAHQDRHGDT ALHAAARQGP DAYTDFFLPL LSRCPSAMGI KNKDGETPGQ ILGWGPPWDS AEEEEEDDAS KEREWRQKLQ GELEDEWQEV MGRFEGDASH ETQEPESFSA WSDRLAREHA QKCQQQQREA EGSRRPPRAE GSSQSWRQQE EEQRLFRERA RAKEEELRES RARRAQEALG DREPKPTRAG PREEHPRGAG RGSLWRFGDV PWPCPGGGDP EAMAAALVAR GPPLEEQGAL RRYLRVQQVR WHPDRFLQRF RSQIETWELG RVMGAVTALS QALNRHAEAL K; Positive Samples: U-87MG, HL-60, HepG2; Cellular Location: Nucleus.

NF-kappa-B inhibitor-like protein 1 (NFKBIL1), also named Ikb like and IkbL1, is a 381 amino acid member of the inhibitor of NF-kappaB (IkappaB) family of proteins. NFKBIL1 contains two ANK repeats and is expressed in the rheumatoid synovial lining and sub-lining layers and co-localized in CD68+ and CD3+, but not Factor VIII+ cells. It is reported that defects in NFKBIL1 expression causes of susceptibility to rheumatoid arthritis (RA), which is a systemic inflammatory disease with autoimmune features and affects the joints. Rheumatoid arthritis (RA) is characterized by inflammatory changes in the synovial membranes and articular structures, widespread fibrinoid degeneration of the collagen fibers in mesenchymal tissues, and by atrophy and rarefaction of bony structures.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFKBIL1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NFKBIL1
Gene Accession No. Q9UBC1
Gene Alias DADB-70P7.2; Def-7; ikappaBL; I-kappa-B-like protein; Ikbl; Inhibitor of kappa B-like protein; LST1; NF-kappa-B inhibitor-like protein 1; NFKB inhibitor like 1; NFKBIL; NFKBIL1; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor like 1; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 1
Gene Symbols NFKBIL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-381 of human NFKBIL1 (NP_0049983).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4795
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.