Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFKBIL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2841780.1ML
Description
NFKBIL1 Polyclonal specifically detects NFKBIL1 in Human samples. It is validated for Western Blot.Specifications
NFKBIL1 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
4795 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
IkappaBL, I-kappa-B-like protein, IKBLikappaBL, Inhibitor of kappa B-like protein, LST1, NF-kappa-B inhibitor-like protein 1, nuclear factor of kappa light polypeptide gene enhancer in B-cellsinhibitor-like 1NFKBIL | |
The immunogen is a synthetic peptide directed towards the N terminal region of human NFKBIL1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction