Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFKBIL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561245
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561245 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561245 Supplier Invitrogen™ Supplier No. PA561245
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IRVRVQVQDH LFLIPVPHSS DTHSVAWLAE QAAQRYYQTC GLLPRLTLRK EGALLAPQDL IPDVLQSNDE VLAEVTSWDL PPLTDRYRRA CQSLGQGEHQ QVLQAVELQG LGLSFSACSL ALDQAQLTPL LRALKLHT Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 88%.

The protein encoded by this gene is thought to be a negative regulator of NF-kappa-B mediated transcription. The encoded protein may bind NF-kappa-B complexes and trap them in the cytoplasm, preventing them from entering the nucleus and interacting with the DNA. Phosphorylation of this protein targets it for degradation by the ubiquitination pathway, which frees the NF-kappa-B complexes to enter the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFKBIL2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TONSL
Gene Accession No. Q96HA7
Gene Alias DNA repair protein; ikappaBR; I-kappa-B-related protein; IKBR; Inhibitor of kappa B-related protein; NF-kappa-B inhibitor-like protein 2; NFKBIL2; Nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 2; TONSL; tonsoku-like; Tonsoku-like protein; tonsoku-like, DNA repair protein
Gene Symbols TONSL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NFKBIL2. Recombinant protein control fragment (Product #RP-103689).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4796
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.