Learn More
NFKBIL2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303843A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is thought to be a negative regulator of NF-kappa-B mediated transcription. The encoded protein may bind NF-kappa-B complexes and trap them in the cytoplasm, preventing them from entering the nucleus and interacting with the DNA. Phosphorylation of this protein targets it for degradation by the ubiquitination pathway, which frees the NF-kappa-B complexes to enter the nucleus.Specifications
NFKBIL2 | |
Polyclonal | |
Unconjugated | |
TONSL | |
IKBR, NFKBIL2, I-kappa-B-related protein, NF-kappa-B inhibitor-like protein 2, ikappaBR, inhibitor of kappa B-related protein, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 2, tonsoku-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4796 | |
4° C | |
Liquid |
Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q96HA7 | |
TONSL | |
Between 425 and 475 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.