Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs NFPA Placard Kits
Click to view available options
Dimensions (L x W):
10 x 10 in.
15 x 15 in.
Description
Includes
One NFPA placard and a full set of self-adhering number panels
- Ideal for mounting on or near containers of hazardous materials and equipment
- Blank panels printed with the proper color coding using UV stable inks
- Hazard panels printed on 4mil clear, self-adhesive polyester
- Numbers and specific hazard symbols included (3 sets of 0-4 and six symbols)
- Choice of placards made of either durable pressure-sensitive vinyl that mounts on most clean, dry surfaces, or high-gloss, 0.060 in. Dura-Plastic suitable for indoor/outdoor use
Specifications
Specifications
Includes | 1 NFPA placard and a full set of self-adhering number panels |
Material | Vinyl |
Quantity | 1 Ea. |
For Use With (Application) | Mounting on or near containers of hazardous materials and equipment |
Height (English) | 10 in. |
Max. Temperature (English) | 176°F |
Min. Temperature (English) | -20°F |
Width (English) | 10 in. |
Temperature Range (Metric) | -28°C to +80°C |
Temperature Range (English) | -20°F to +176°F |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction