Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFRKB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552389
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552389 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552389 Supplier Invitrogen™ Supplier No. PA552389
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSSSGVLLVS SPTMPHLGTM LSPASSQTAP SSQAAARVVS HSGSAGLSQV RVVAQPSLPA VPQQSGGPAQ TLPQMPAGPQ IRVPATATQT KVVPQTVMAT VPVKAQTTAA TV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

NFRKB (nuclear factor related to kappa-B-binding protein) was first identified as a factor that could bind a variant NF-kappa B site in the interleukin-2 receptor alpha (IL-2R alpha) gene. NFRKB appears to be a transcription factor central to immune cell development. Evidence of this comes from the identification of NFRKB as an amplified gene in several cases of acute myeloid leukemia. Additionally, in another clinical case, a partial deletion of the chromosomal region that includes NFRKB is shown to be associated with combined cellular immunodeficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFRKB
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NFRKB
Gene Accession No. Q6P4R8
Gene Alias A530090G11Rik; DNA-binding protein R kappa B; DNA-binding protein R kappa-B; INO80 complex subunit G; INO80G; NFRKB; nuclear factor related to kappa B binding protein; nuclear factor related to kappaB binding protein; nuclear factor related to kappa-B-binding protein
Gene Symbols NFRKB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NFRKB. Recombinant protein control fragment (Product #RP-89400).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4798
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.