Learn More
NFRKB Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301460A
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Binds to the DNA consensus sequence 5′-GGGGAATCTCC-3′. Modulates the deubiquitinase activity of UCHL5.Specifications
| NFRKB | |
| Polyclonal | |
| Unconjugated | |
| NFRKB | |
| INO80G, DNA-binding protein R kappa B, DNA-binding protein R kappa-B, INO80 complex subunit G, nuclear factor related to kappa-B-binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4798 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q6P4R8 | |
| NFRKB | |
| Between 1249 and 1299 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.