Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23793325UL
Description
NFS1 Polyclonal specifically detects NFS1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NFS1 | |
Polyclonal | |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Q9Y697 | |
NFS1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: GVGAIYIRRRPRVRVEALQSGGGQERGMRSGTVPTPLVVGLGAACEVAQQEMEYDHKRISKLSERLIQNIMKSLPDVVMNGDPKHHYPGCINLSF | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
cysteine desulfurase, mitochondrial, EC 2.8.1.7, NFS1 nitrogen fixation 1 homolog (S. cerevisiae), NifS, NIFShomolog), nitrogen-fixing bacteria S-like protein | |
Rabbit | |
Affinity Purified | |
RUO | |
9054 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction