Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NFS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562485 Supplier Invitrogen™ Supplier No. PA562485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVGAIYIRRR PRVRVEALQS GGGQERGMRS GTVPTPLVVG LGAACEVAQQ EMEYDHKRIS KLSERLIQNI MKSLPDVVMN GDPKHHYPGC INLSF Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

NFS1 (nitrogen fixation 1), also known as NIFS or IscS (cysteine desulfurase), is a member of the class V pyridoxal-phosphate-dependent aminotransferase family. It localizes to the cytoplasm or mitochondrion depending on which form is generated based on cytosolic pH. Highest expression levels of NFS1 are found in heart and skeletal muscle. Lower levels of expression are also found in liver, brain and pancreas. NFS1 is responsible for catalyzing the removal of sulfur from cysteine to form alanine, thereby supplying the inorganic sulfur for iron-sulfur (Fe-S) clusters. Fe-S clusters function as essential cofactors in a wide variety of events, including facilitation of electron transfer processes in oxidative phosphorylation, catalysis of enzymatic reactions in aconitase and dehydratases and maintenance of structural integrity in the DNA repair enzyme endonuclease III.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFS1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NFS1
Gene Accession No. Q9Y697
Gene Alias AA987187; cysteine desulfurase, mitochondrial; Cysteine desulfurase, mitochondrial-like protein; HUSSY-08; IscS; LOW QUALITY PROTEIN: cysteine desulfurase, mitochondrial; m-Nfs1; m-Nfsl; Nfs1; NFS1 cysteine desulfurase; NFS1 nitrogen fixation 1 homolog; NFS1 nitrogen fixation 1 homolog (S. cerevisiae); NFS1, cysteine desulfurase; Nifs; nifS-like (sic); nitrogen fixation 1 (S. cerevisiae, homolog); nitrogen fixation gene 1; nitrogen fixation gene 1 (S. cerevisiae); nitrogen fixation gene, yeast homolog 1; nitrogen-fixing bacteria S-like protein
Gene Symbols NFS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NFS1. Recombinant protein control fragment (Product #RP-103109).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9054
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.